siRNA Details
Gene Symbol: Fundc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026126.4 | NP_080402.3 | 2 | 90 | ||
GenBank mRNA | AK013460.1 | - | 2 | 68 | ||
AK014705.1 | - | 2 | 67 | |||
AK031214.1 | - | 2 | 67 | |||
AY234858.1 | - | 2 | 56 | |||
BC042794.1 | - | 1 | 47 | |||
BC049566.1 | - | 2 | 65 | 6742356 | ||
BC061464.1 | - | 2 | 60 | 6819539 | ||
BY124891.1 | - | 2 | 90 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
