siRNA Details
Gene Symbol: Ppp1r2-ps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015065.1 | - | 1 | 271 | ||
AK015311.1 | - | 1 | 243 | |||
BC100448.1 | - | 1 | 243 | 30918995 | ||
BC132484.1 | - | 1 | 215 | 40130761 | ||
BC132486.1 | - | 1 | 215 | 40130763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
