siRNA Details
Gene Symbol: Srpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026130.1 | NP_080406.1 | 3 | 373 | ||
GenBank mRNA | AK004967.2 | - | 3 | 373 | ||
AK007516.1 | - | 3 | 398 | |||
AK038017.1 | - | 3 | 361 | |||
BC021839.1 | - | 3 | 356 | 4207633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
