siRNA Details
Gene Symbol: Pdlim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114087.2 | NP_001107559.1 | 7, 8 | 671 | ||
NM_026131.4 | NP_080407.3 | 7, 8 | 569 | |||
GenBank mRNA | AK003019.1 | - | 8, 9 | 645 | ||
AK003338.1 | - | 7, 8 | 560 | |||
AK010141.1 | - | 7, 8 | 558 | |||
AK077998.1 | - | 7, 8 | 663 | |||
BC049565.1 | - | 7, 8 | 557 | 6588160 | ||
BC052698.1 | - | 7, 8 | 642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
