siRNA Details
Gene Symbol: Far1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285831.1 | NP_001272760.1 | 13 | 1646 | ||
NM_027379.3 | NP_081655.2 | 13 | 1646 | |||
GenBank mRNA | AK005531.1 | - | 13 | 1620 | ||
AK014405.1 | - | 13 | 1618 | |||
AK014486.1 | - | 13 | 1619 | |||
AK030067.1 | - | 13 | 1577 | |||
AK033674.1 | - | 13 | 1589 | |||
BC007178.1 | - | 13 | 1594 | 3495305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
