siRNA Details
Gene Symbol: Rps20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026147.5 | NP_080423.1 | 4 | 400 | ||
GenBank mRNA | AK002405.1 | - | 4 | 401 | ||
AK014594.1 | - | 400 | ||||
AK136398.1 | - | 4 | 403 | |||
AK145656.1 | - | 4 | 401 | |||
AK150867.1 | - | 4 | 400 | |||
AK150972.1 | - | 4 | 400 | |||
BC011323.1 | - | 4 | 387 | 3257215 | ||
BC090389.1 | - | 4 | 391 | 3482252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
