siRNA Details
Gene Symbol: 4921536K21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026150.3 | NP_080426.1 | 2 | 307 | ||
GenBank mRNA | AK006513.1 | - | 2 | 280 | ||
AK015005.1 | - | 2 | 280 | |||
AK049235.1 | - | 2 | 279 | |||
AK162162.1 | - | 2 | 165 | |||
BC048445.1 | - | 2 | 276 | 6704927 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
