siRNA Details
Gene Symbol: Ccdc127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168658.1 | NP_001162129.1 | 2 | 182 | ||
NM_001168659.1 | NP_001162130.1 | 2 | 182 | |||
NM_024201.3 | NP_077163.1 | 2 | 176 | |||
GenBank mRNA | AK002564.1 | - | 2 | 164 | ||
AK007738.1 | - | 2 | 128 | |||
AK164539.1 | - | 2 | 183 | |||
AK170932.1 | - | 2 | 169 | |||
BC003345.1 | - | 2 | 156 | 3257525 | ||
BC049099.1 | - | 2 | 134 | 5721356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
