siRNA Details
Gene Symbol: Map1lc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026160.4 | NP_080436.1 | 2, 3 | 154 | ||
GenBank mRNA | AF255953.1 | - | 2, 3 | 172 | ||
AK002795.1 | - | 2, 3 | 157 | |||
AK003106.1 | - | 2, 3 | 155 | |||
AK003205.1 | - | 2, 3 | 155 | |||
AK003558.1 | - | 2, 3 | 157 | |||
AK012604.1 | - | 2, 3 | 158 | |||
AK132329.1 | - | 162 | ||||
AK151614.1 | - | 2, 3 | 153 | |||
BC068180.1 | - | 2, 3 | 157 | 30634082 | ||
BC110308.1 | - | 2, 3 | 145 | 5319360 |

More Information
Other Species Targeted

Gene Ontology Categories:
