siRNA Details
Gene Symbol: Ikbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026166.2 | NP_080442.2 | 2 | 290 | ||
NM_027078.2 | NP_081354.1 | 2 | 290 | |||
GenBank mRNA | AK004670.1 | - | 2 | 248 | ||
AK006283.1 | - | 2 | 258 | |||
BC034890.1 | - | 2 | 252 | 1447362 | ||
BC055834.1 | - | 2 | 241 | 3492302 | ||
CF171082.1 | - | 2 | 272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
