siRNA Details
Gene Symbol: Snap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023348.4 | NP_075837.3 | 5 | 863 | ||
GenBank mRNA | AK005042.1 | - | 5 | 813 | ||
AK007065.1 | - | 5 | 856 | |||
AK039203.1 | - | 5 | 832 | |||
AK147845.1 | - | 5 | 771 | |||
AY008722.1 | - | 5 | 860 | |||
BC030066.1 | - | 5 | 812 | 5118167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
