siRNA Details
Gene Symbol: 1700028P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026188.2 | NP_080464.1 | 4, 5 | 540 | ||
GenBank mRNA | AK006473.1 | - | 4, 5 | 543 | ||
AK014972.1 | - | 4, 5 | 542 | |||
BC049563.1 | - | 4, 5 | 525 | 6771302 |
