siRNA Details
Gene Symbol: Tmem167b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026198.2 | NP_080474.2 | 2 | 227 | ||
GenBank mRNA | AK008415.1 | - | 2 | 140 | ||
AK046697.1 | - | 2 | 128 | |||
BC050932.1 | - | 2 | 131 | 6811263 | ||
BC055449.1 | - | 2 | 144 | 3993993 | ||
BQ560740.1 | - | 2 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
