siRNA Details
Gene Symbol: Kcnv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026200.3 | NP_080476.2 | 2 | 802 | ||
GenBank mRNA | AK012275.1 | - | 2 | 597 | ||
AK034091.1 | - | 2 | 695 | |||
AK038450.1 | - | 2 | 789 | |||
AK076120.1 | - | 2 | 571 | |||
AK139482.1 | - | 2 | 638 | |||
BC117819.1 | - | 2 | 691 | |||
BC128477.1 | - | 2 | 691 | 40092817 | ||
BC128478.1 | - | 2 | 691 | 40092823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
