siRNA Details
Gene Symbol: Rnf151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026205.3 | NP_080481.1 | 3 | 271 | ||
GenBank mRNA | AK005850.1 | - | 3 | 232 | ||
AK005854.1 | - | 3 | 227 | |||
BC049562.1 | - | 3 | 271 | 6774166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
