siRNA Details
Gene Symbol: Kctd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026214.3 | NP_080490.3 | 2 | 1010 | ||
GenBank mRNA | AK008741.1 | - | 2 | 1005 | ||
AK017592.1 | - | 1 | 741 | |||
AK032314.1 | - | 1 | 717 | |||
AK032624.1 | - | 1 | 748 | |||
BC028748.1 | - | 1 | 725 | 1332429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
