siRNA Details
Gene Symbol: Fgfr1op2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026218.2 | NP_080494.1 | 3 | 519 | ||
GenBank mRNA | AK004662.1 | - | 3 | 503 | ||
AK005320.1 | - | 3 | 496 | |||
AK008983.1 | - | 3 | 503 | |||
AK050102.1 | - | 3 | 519 | |||
AK151318.1 | - | 3 | 512 | |||
AK152617.1 | - | 3 | 512 | |||
AK153137.1 | - | 3 | 512 | |||
AK159805.1 | - | 3 | 468 | |||
AK169380.1 | - | 3 | 528 | |||
AK169383.1 | - | 3 | 496 | |||
BC028876.1 | - | 3 | 482 | 4504364 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_201421, XM_008763402, XM_006237688, AY426740, AY426739, and others.
Gene Ontology Categories:
