siRNA Details
Gene Symbol: Ppfibp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170433.1 | NP_001163904.1 | 26 | 3083 | ||
NM_026221.2 | NP_080497.1 | 25 | 3050 | |||
GenBank mRNA | AK014559.1 | - | 9 | 1216 | ||
AK146575.1 | - | 25 | 3034 | |||
AK154482.1 | - | 25 | 2995 | |||
AK173133.1 | - | 26 | 2988 | |||
BC035209.1 | - | 25 | 2896 | |||
BC049862.1 | - | 4 | 544 | |||
BC058176.1 | - | 13 | 1586 | |||
BC060078.1 | - | 16 | 3323 | |||
BC071257.1 | - | 6 | 758 | |||
BC072603.1 | - | 25 | 3030 | 6830850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
