siRNA Details
Gene Symbol: Ttll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014974.1 | NP_001014974.1 | 17, 18 | 3587 | ||
GenBank mRNA | AK129075.1 | - | 15, 16 | 2414 | ||
AM690747.1 | - | 15, 16 | 3190 | |||
BC044790.1 | - | 4, 5 | 564 | |||
BC049820.1 | - | 2, 3 | 818 | |||
BC085151.1 | - | 17, 18 | 3587 | 30534216 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
