siRNA Details
Gene Symbol: Ttll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014974.1 | NP_001014974.1 | 11 | 2723 | ||
GenBank mRNA | AK129075.1 | - | 9 | 1550 | ||
AM690747.1 | - | 9 | 2326 | |||
BC085151.1 | - | 11 | 2723 | 30534216 |

More Information
Other Species Targeted

Gene Ontology Categories:
