siRNA Details
Gene Symbol: Ttll4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014974.1 | NP_001014974.1 | 12 | 2816 | ||
GenBank mRNA | AK129075.1 | - | 10 | 1643 | ||
AM690747.1 | - | 10 | 2419 | |||
BC085151.1 | - | 12 | 2816 | 30534216 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_001074653.
Gene Ontology Categories:
