siRNA Details
Gene Symbol: Glipr1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026223.2 | NP_080499.1 | 1 | 66 | ||
GenBank mRNA | AK014846.1 | - | 1 | 67 | ||
AK015834.1 | - | 1 | 50 | |||
AK161292.1 | - | 1 | 67 | |||
BC118017.1 | - | 1 | 67 | 40102576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
