siRNA Details
Gene Symbol: Zswim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178375.2 | NP_848462.1 | 2 | 417 | ||
GenBank mRNA | AK014904.1 | - | 2 | 437 | ||
BC023287.1 | - | 2 | 417 | 3993352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
