siRNA Details
Gene Symbol: Gpr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026229.4 | NP_080505.1 | 4 | 367 | ||
GenBank mRNA | AK002937.1 | - | 4 | 387 | ||
AK009734.1 | - | 3 | 246 | |||
AK016787.1 | - | 4 | 367 | |||
BC010729.1 | - | 4 | 360 | 3708143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
