siRNA Details
Gene Symbol: Larp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026235.4 | NP_080511.2 | 3 | 968 | ||
GenBank mRNA | AK017372.1 | - | 3 | 973 | ||
AK035809.1 | - | 545 | ||||
AK039614.1 | - | 3 | 951 | |||
AK042616.1 | - | 3 | 900 | |||
AK047452.1 | - | 1 | 272 | |||
AK087521.1 | - | 3 | 971 | |||
BC090615.1 | - | 3 | 1100 | 6809572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
