siRNA Details
Gene Symbol: Tmem35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026239.2 | NP_080515.1 | 2 | 487 | ||
GenBank mRNA | AK005180.1 | - | 2 | 322 | ||
AK018535.1 | - | 2 | 324 | |||
AK042538.1 | - | 2 | 323 | |||
AK043881.1 | - | 2 | 322 | |||
BC042623.1 | - | 2 | 487 | 5369317 | ||
BC050880.1 | - | 2 | 389 | 5690508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
