siRNA Details
Gene Symbol: Alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026247.3 | NP_080523.2 | 3, 4 | 467 | ||
GenBank mRNA | AK008162.1 | - | 3, 4 | 417 | ||
AK012910.1 | - | 479 | ||||
AK014790.1 | - | 3, 4 | 412 | |||
AK086546.1 | - | 3, 4 | 360 | |||
AK089102.1 | - | 3, 4 | 431 | |||
BC137692.1 | - | 3, 4 | 467 | 8860711 |

More Information
Other Species Targeted

Gene Ontology Categories:
