siRNA Details
Gene Symbol: Alg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026247.3 | NP_080523.2 | 2 | 269 | ||
GenBank mRNA | AK008162.1 | - | 2 | 219 | ||
AK012910.1 | - | 281 | ||||
AK014790.1 | - | 2 | 214 | |||
AK086546.1 | - | 2 | 162 | |||
AK089102.1 | - | 2 | 233 | |||
BC053004.1 | - | 2 | 206 | |||
BC137692.1 | - | 2 | 269 | 8860711 |
