siRNA Details
Gene Symbol: Lrrc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146021.1 | NP_001139493.1 | 3 | 537 | ||
NM_026253.4 | NP_080529.1 | 3 | 537 | |||
GenBank mRNA | AK005666.1 | - | 3 | 477 | ||
AK006063.1 | - | 3 | 474 | |||
AK014952.1 | - | 3 | 502 | |||
AK015365.1 | - | 3 | 541 | |||
AK029909.1 | - | 3 | 502 | |||
AY775400.1 | - | 3 | 606 | |||
BC048444.1 | - | 3 | 522 | |||
BC061072.1 | - | 1 | 265 | 6771444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
