siRNA Details
Gene Symbol: 4930524B15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026262.1 | NP_080538.1 | 2 | 335 | ||
GenBank mRNA | AK015006.1 | - | 2 | 377 | ||
AK015879.1 | - | 2 | 335 | |||
BC114574.1 | - | 2 | 325 | 40049349 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
