siRNA Details
Gene Symbol: 4930519G04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026263.2 | NP_080539.1 | 5 | 473 | ||
GenBank mRNA | AK015843.1 | - | 5 | 475 | ||
AK015857.1 | - | 5 | 524 | |||
AK161377.1 | - | 3 | 1374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
