siRNA Details
Gene Symbol: Necap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026267.2 | NP_080543.2 | 6, 7 | 694 | ||
GenBank mRNA | AK004805.1 | - | 6, 7 | 695 | ||
AK005263.1 | - | 6, 7 | 685 | |||
AK081598.1 | - | 6, 7 | 695 | |||
AK168500.1 | - | 6, 7 | 695 | |||
AK212285.1 | - | 1, 2 | 117 | |||
AK220236.1 | - | 6, 7 | 680 | |||
BC011466.1 | - | 6, 7 | 693 | 4013043 | ||
BC119133.1 | - | 6, 7 | 706 | 8733882 | ||
BC120841.1 | - | 6, 7 | 777 | 40129764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
