siRNA Details
Gene Symbol: Fibin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026271.1 | NP_080547.1 | 1 | 317 | ||
GenBank mRNA | AB236893.1 | - | 1 | 119 | ||
AK003790.1 | - | 1 | 317 | |||
AK011548.1 | - | 1 | 488 | |||
AK012026.1 | - | 1 | 500 | |||
AK159648.1 | - | 1 | 450 | |||
BC027250.1 | - | 1 | 306 | 3603588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
