siRNA Details
Gene Symbol: Spc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026282.5 | NP_080558.1 | 4 | 440 | ||
GenBank mRNA | AK011728.1 | - | 4 | 458 | ||
AK031680.1 | - | 3 | 533 | |||
AK075950.1 | - | 4 | 441 | |||
AK153185.1 | - | 4 | 458 | |||
BC086683.1 | - | 4 | 443 | 30676712 | ||
BC098472.1 | - | 448 | ||||
BC116238.1 | - | 4 | 441 | 40091783 | ||
BC116239.1 | - | 4 | 440 | 40091784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
