siRNA Details
Gene Symbol: Lyrm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163628.1 | NP_001157100.1 | 3 | 557 | ||
NM_133688.3 | NP_598449.1 | 3 | 320 | |||
GenBank mRNA | AF412298.1 | - | 3 | 974 | ||
AF412299.1 | - | 4 | 555 | |||
AF412300.1 | - | 3 | 321 | |||
AK015530.1 | - | 3 | 293 | |||
AK075835.1 | - | 3 | 301 | |||
AY753325.1 | - | 3 | 312 | |||
BC021522.1 | - | 3 | 268 | 5352723 | ||
CX203242.1 | - | 3 | 557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
