siRNA Details
Gene Symbol: Rabl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042499.1 | NP_001035964.1 | 3 | 268 | ||
GenBank mRNA | AK011280.1 | - | 3 | 263 | ||
AK016099.1 | - | 3 | 277 | |||
AK028195.1 | - | 3 | 262 | |||
AK028269.1 | - | 3 | 255 | |||
AK156982.1 | - | 3 | 259 | |||
BC050194.1 | - | 3 | 249 | 30022531 |

More Information
Other Species Targeted

Gene Ontology Categories:
