siRNA Details
Gene Symbol: Rpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026308.2 | NP_080584.1 | 4 | 314 | ||
GenBank mRNA | AJ504717.1 | - | 4 | 321 | ||
AK002780.1 | - | 4 | 315 | |||
AK003455.1 | - | 4 | 312 | |||
AY251016.1 | - | 4 | 336 | |||
AY251017.1 | - | 3 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
