siRNA Details
Gene Symbol: Lsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026309.2 | NP_080585.1 | 3 | 216 | ||
GenBank mRNA | AK003125.1 | - | 3 | 219 | ||
AK007849.1 | - | 3 | 208 | |||
AK021251.1 | - | 3 | 192 | |||
BC054368.1 | - | 3 | 204 | 5353718 | ||
BC062875.1 | - | 3 | 250 | 5684590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
