siRNA Details
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023374.3 | NP_075863.2 | 5 | 589 | ||
GenBank mRNA | AF095936.1 | - | 2 | 140 | ||
AK003052.1 | - | 5 | 483 | |||
AK003533.1 | - | 5 | 473 | |||
AK009660.1 | - | 5 | 483 | |||
AK153045.1 | - | 5 | 466 | |||
AK169252.1 | - | 5 | 480 | |||
AK169839.1 | - | 5 | 482 | |||
AK190493.1 | - | 2 | 61 | |||
BC013509.1 | - | 5 | 484 | 4225025 | ||
BC018580.1 | - | 5 | 448 | |||
BC051934.1 | - | 5 | 589 | 5688898 | ||
DQ403000.1 | - | 4 | 380 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
