siRNA Details
Gene Symbol: Sdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023374.3 | NP_075863.2 | 3 | 357 | ||
GenBank mRNA | AK003052.1 | - | 3 | 251 | ||
AK003533.1 | - | 3 | 241 | |||
AK009660.1 | - | 3 | 251 | |||
AK153045.1 | - | 3 | 234 | |||
AK169252.1 | - | 3 | 248 | |||
AK169839.1 | - | 3 | 250 | |||
AK196072.1 | - | 1 | 14 | |||
BC013509.1 | - | 3 | 252 | 4225025 | ||
BC018580.1 | - | 3 | 216 | |||
BC051934.1 | - | 3 | 357 | 5688898 | ||
DQ403000.1 | - | 2 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
