siRNA Details
Gene Symbol: Ost4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134692.2 | NP_001128164.1 | 2 | 487 | ||
NM_024460.4 | NP_077780.3 | 3 | 359 | |||
GenBank mRNA | AA563474.1 | - | 3 | 359 | ||
AK002224.2 | - | 3 | 334 | |||
AK002453.1 | - | 3 | 281 | |||
AK003569.1 | - | 3 | 338 | |||
AK005413.1 | - | 3 | 282 | |||
AK009384.1 | - | 294 | ||||
AK010297.1 | - | 2 | 423 | |||
AK019285.1 | - | 3 | 319 | |||
BC002177.1 | - | 2 | 424 | 3486998 | ||
BC082316.1 | - | 2 | 398 | 6814413 | ||
CA466367.1 | - | 3 | 317 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134690, NM_001134691, CB706964, BC157814.
Gene Ontology Categories:
