siRNA Details
Gene Symbol: Fam174a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026321.4 | NP_080597.2 | 2 | 639 | ||
GenBank mRNA | AK009892.1 | - | 2 | 578 | ||
AK017318.1 | - | 2 | 612 | |||
AK033948.1 | - | 2 | 524 | |||
AK084861.1 | - | 2 | 640 | |||
AK148807.1 | - | 2 | 540 | |||
BC021428.1 | - | 2 | 654 | 5034950 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
