siRNA Details
Gene Symbol: 1810037I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024461.2 | NP_077781.1 | 2 | 693 | ||
GenBank mRNA | AK170776.1 | - | 2 | 659 | ||
BC091739.1 | - | 2 | 661 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1810037I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024461.2 | NP_077781.1 | 2 | 693 | ||
GenBank mRNA | AK170776.1 | - | 2 | 659 | ||
BC091739.1 | - | 2 | 661 |