siRNA Details
Gene Symbol: 1810037I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024461.2 | NP_077781.1 | 1 | 172 | ||
GenBank mRNA | AK007721.1 | - | 1 | 173 | ||
AK009748.1 | - | 1 | 138 | |||
AK013091.1 | - | 1 | 157 | |||
AK170776.1 | - | 1 | 138 | |||
BC002135.1 | - | 1 | 181 | 3484100 | ||
BC086913.1 | - | 1 | 137 | 30121587 | ||
BC091739.1 | - | 1 | 140 |
