siRNA Details
Gene Symbol: Dph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026344.3 | NP_080620.1 | 1 | 257 | ||
GenBank mRNA | AK011199.1 | - | 1 | 275 | ||
AK018638.1 | - | 1 | 258 | |||
AK051026.1 | - | 1 | 276 | |||
AK080747.1 | - | 1 | 275 | |||
BC014746.1 | - | 1 | 277 | |||
BC031124.1 | - | 1 | 244 | 5029876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
