siRNA Details
Gene Symbol: Fbxo32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026346.3 | NP_080622.1 | 6 | 780 | ||
GenBank mRNA | AF441120.1 | - | 6 | 826 | ||
AK014612.1 | - | 6 | 733 | |||
AK014793.1 | - | 6 | 762 | |||
BC027211.1 | - | 6 | 714 | 3981597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
