siRNA Details
Gene Symbol: Itgb3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026348.3 | NP_080624.1 | 3 | 206 | ||
GenBank mRNA | AK005779.1 | - | 3 | 275 | ||
AK013373.1 | - | 3 | 221 | |||
AK015557.1 | - | 3 | 231 | |||
AK045037.1 | - | 3 | 207 | |||
AK166362.1 | - | 3 | 207 | |||
BC049557.1 | - | 3 | 216 | 6741650 | ||
EF437329.1 | - | 1 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
