siRNA Details
Gene Symbol: 4930579F01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163385.1 | NP_001156857.1 | 2 | 637 | ||
NM_001163386.1 | NP_001156858.1 | 2 | 520 | |||
GenBank mRNA | AK016318.1 | - | 2 | 520 | ||
BC150693.1 | - | 2 | 597 | 9087604 |
