siRNA Details
Gene Symbol: Arpc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026369.2 | NP_080645.2 | 1 | 229 | ||
GenBank mRNA | AK008097.1 | - | 1 | 120 | ||
AK011348.1 | - | 1 | 106 | |||
AK011911.1 | - | 1 | 106 | |||
AK017669.1 | - | 1 | 107 | |||
AK017985.1 | - | 1 | 140 | |||
AK019149.1 | - | 1 | 106 | |||
AK087427.1 | - | 1 | 96 | |||
AK149756.1 | - | 1 | 98 | |||
AK150443.1 | - | 1 | 106 | |||
AK150703.1 | - | 1 | 106 | |||
AK151490.1 | - | 1 | 91 | |||
AK151654.1 | - | 1 | 97 | |||
AK151656.1 | - | 1 | 119 | |||
AK152281.1 | - | 1 | 119 | |||
AK153262.1 | - | 1 | 100 | |||
AK159407.1 | - | 1 | 153 | |||
AK159438.1 | - | 1 | 93 | |||
AK159738.1 | - | 1 | 116 | |||
AK166284.1 | - | 1 | 121 | |||
AK166335.1 | - | 1 | 136 | |||
AK166725.1 | - | 1 | 120 | |||
AK166919.1 | - | 1 | 122 | |||
AK167916.1 | - | 1 | 90 | |||
AK168264.1 | - | 1 | 97 | |||
AK168986.1 | - | 1 | 68 | |||
BC060143.1 | - | 1 | 135 | 6411792 | ||
CF170085.1 | - | 1 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
