siRNA Details
Gene Symbol: Rtp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023386.5 | NP_075875.3 | 2 | 299 | ||
GenBank mRNA | AJ251364.1 | - | 2 | 294 | ||
AK007477.1 | - | 2 | 277 | |||
AK018021.1 | - | 2 | 292 | |||
AY562228.1 | - | 2 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
